Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ DC1500 Chlorine Colorimeter Kit - DPD Liquid
SDPSCI_ED

Catalog No. S28426 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Chlorine Colorimeter Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S28426 Chlorine Colorimeter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28426 Supplier LaMotte™ Supplier No. 3240LI
Add to Cart
Add to Cart

Includes

1500 Colorimeter for Chlorine DPD, 30mL DPD 1A Free Chlorine Reagent, 30mL DPD 1B Free Chlorine Reagent, 30mL DPD 3 Total Chlorine Reagent, set of six colorimeter tubes with caps, water sample collecting bottle, USB wall adapter, USB Cable, Manual, Quick Start Guide, hard-shell carry case.

Kit covers entire critical chlorine range of 0 to 4 ppm with an MDL of 0.03 ppm

  • Large graphical backlit LCD display
  • Simple, menu-driven operation
  • IP67 Waterproof
  • Auto-off (disabled, 5, 10, 50 minutes)
  • LED (four) light source
  • Four silicon photodiode with integrated interference detectors
  • Mini USB port for output
  • Three modes: pre-programmed test, absorbance, %T
  • Sample chamber accepts 25 mm diameter flat bottom test tubes
  • Can be interfaced with any Windows-based computer by using the LaMotte SMARTLink™ 3 Program and USB Cable
  • Ideal for both field and lab use
  • Specifications:
    • Readout: 160 x 100 backlit LCD, 20 x 6 line graphical
    • Wavelengths 525nm (Abs 428nm, 525nm, 568nm, 635nm)
    • Wavelength Accuracy: ±2% FS
    • Wavelength Bandwidth: 10nm typical
    • Photometric Range: -2 to +2 AU
    • Languages: English, Spanish, French, Portuguese, Italian
    • Power: Lithium ion rechargeable battery, 3.7V, 2.5 x 0.75
    • Battery: Charge Life: Approx. 380 tests with backlight
    • Battery Life: Approx. 500 charges
  • Applications: Industrial Water Boiling and Cooling, Water and Wastewater, drinking water supplies and distribution systems, swimming pool and spas, sewage and chlorinated waste waters, process waters and sanitizing solutions.

Certifications

European CE marked

Warranty and Services

Two years

Specifications

Absorbance Range 428 nm, 525 nm, 568 nm, 635 nm
Backlight Option Yes
Battery Life Approx. 500 charges
Certifications/Compliance European CE marked
Battery Type Rechargeable Lithium-Ion
Detector Type Silicon Photodiode
Display Backlit LCD
IP Rating IP67
Languages English, French, Spanish, Italian, Portuguese
Light Source 4 x LED
Spectral Bandwidth 10 nm
Type Chlorine Colorimeter Kit
For Use With (Application) Industrial Water Boiling and Cooling, Water and Wastewater, drinking water supplies and distribution systems, swimming pool andspas, sewage and chlorinated waste waters, process waters and sanitizing solutions
Includes 1500 Colorimeter for Chlorine DPD, 30mL DPD 1A Free Chlorine Reagent, 30mL DPD 1B Free Chlorine Reagent, 30mL DPD 3 Total Chlorine Reagent, set of six colorimeter tubes with caps, water sample collecting bottle, USB wall adapter, USB Cable, DC1500-CL Chlorine Colorimeter DPD Liquid Manual, DC1500-CL Chlorine Colorimeter DPD Liquid Quick Start Guide
Measurement Range 0 to 4 ppm with an MDL of 0.03 ppm
Model DC1500
Outputs Mini USB port
Reagent Form Liquid
Software Details LaMotte SMARTLink 3
Test Method Three: pre-programmed test, absorbance, %T
Warranty 2 Years
Wavelength 525 nm
Wavelength Accuracy ± 2% FS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.