Learn More
Medchemexpress LLC HY-111621 10mg Medchemexpress, DC661 CAS:1872387-43-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11162110MG
Encompass_Preferred
Medchemexpress, HY-111621 10mg DC661 CAS:1872387-43-3 DC661 is a potent palmitoyl-protein thioesterase 1 (PPT1) inhibitor, inhibits autophagy, and acts as an anti-lysosomal agent. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8928
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.