Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAA (2,4-Dichlorophenylacetic Acid) Solution, 5000μg/mL in methanol, Ultra Scientific

Catalog No. USPPS1621
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPPS1621 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPPS1621 Supplier Agilent Technologies Supplier No. PPS1621
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19719-28-9 , 67-56-1
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
InChI Key GXMWLJKTGBZMBH-UHFFFAOYSA-N
PubChem CID 88209
IUPAC Name 2-(2,4-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C=C1Cl)Cl)CC(=O)O

Specifications

CAS Max % 99.37, 0.63
Analyte or Component Names Methanol,DCAA
Element Methanol,DCAA
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material DCAA Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.