Learn More
Enzo Life Sciences DCC (human) monoclonal antibody (Axel-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804853C100
Encompass_Preferred
Clone: Axel-1. Isotype: IgG2a λ. Immunogen: Recombinant human DCC fibronectin domain 5 (ALX-522-109). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: Flow Cytometry, FUNC. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-0001
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.