Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DcCoV Nucleoprotein Antibody Alexa Fluor™ Plus 594 - (HKU23-368F), Novus Biologicals™
SDP

Catalog No. RD10130043
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10130043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10130043 Supplier R&D Systems Supplier No. NBP348433AFP594

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

DcCoV Nucleoprotein Polyclonal antibody specifically detects DcCoV Nucleoprotein in Virus samples. It is validated for ELISA,Western Blot

Specifications

Antigen DcCoV Nucleoprotein
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor Plus 594
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Dromedary camel coronavirus Nucleoprotein - (HKU23-368F) (RefSeq#: AHN64796.1; Met1-Ile448).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.