Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518592
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518592 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518592 Supplier Invitrogen™ Supplier No. PA518592
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization. Mutations in this gene have been associated with Reading Disability, also referred to as developmental dyslexia. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCDC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Dcdc2a
Gene Accession No. Q5DU00
Gene Alias AW492955; Dcdc2; Dcdc2a; DFNB66; doublecortin domain containing 2; doublecortin domain containing 2a; doublecortin domain-containing protein 2; HGNC:18141; Kiaa1154; MGC54817; NPHP19; Protein RU2S; RU2; RU2S
Gene Symbols Dcdc2a
Host Species Goat
Immunogen Synthetic peptide sequence (PQPLKRKGKKDSN) corresponding to the internal amino acids of Dcdc2a (aa 271-284).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 195208
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.