Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556787 Supplier Invitrogen™ Supplier No. PA556787
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAPEQVEEIL DHSEQQARPA RVNGGTDEEN GEELQQVNNE LQLVLDKERK SQGAGSGQDE ADVDPQRPPR PEVKITSPEE NENNQQNKDY AAVA Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.

This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization. Mutations in this gene have been associated with Reading Disability, also referred to as developmental dyslexia. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCDC2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.80 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCDC2
Gene Accession No. Q9UHG0
Gene Alias AW492955; Dcdc2; Dcdc2a; DFNB66; doublecortin domain containing 2; doublecortin domain containing 2a; doublecortin domain-containing protein 2; HGNC:18141; Kiaa1154; MGC54817; NPHP19; Protein RU2S; RU2; RU2S
Gene Symbols DCDC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCDC2. Recombinant protein control fragment (Product #RP-102596).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51473
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.