Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCDC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546005 Supplier Invitrogen™ Supplier No. PA546005
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KTTEPYAPVR LRVLQNGEKN KNRSVTILGP DISPGRKTQC TEILNLPSAA Sequence homology: Human: 100%; Rabbit: 82%.

This gene encodes a member of the doublecortin family. The protein encoded by this gene is a hydrophilic, intracellular protein. It contains a single doublecortin domain and is unable to bind microtubules and to regulate microtubule polymerization. This gene is mainly expressed in adult testis. It does not have a mouse homolog.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCDC5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DCDC1
Gene Accession No. M0R2J8
Gene Alias DCDC1; DCDC5; doublecortin domain containing 1; Doublecortin domain-containing 5 protein; Doublecortin domain-containing protein 1; KIAA1493
Gene Symbols DCDC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DCDC5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341019
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.