Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCI Recombinant Rabbit Monoclonal Antibody (K01_1Q28)
GREENER_CHOICE

Catalog No. p-200113799
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA563660 100 μL
MA565119 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA563660 Supplier Invitrogen™ Supplier No. MA563660
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DADVQNFVSF ISKD.

Protein names: Enoyl-CoA delta isomerase 1, mitochondrial; 3,2-trans-enoyl-CoA isomerase; Delta(3),Delta(2)-enoyl-CoA isomerase; Short name=D3,D2-enoyl-CoA isomerase; Dodecenoyl-CoA isomerase. Gene names: ECI1, DCI. Function: Able to isomerize both 3-cis and 3-trans double bonds into the 2-trans form in a range of enoyl-CoA species. Catalytic activity: (3Z)-dodec-3-enoyl-CoA = (2E)-dodec-2-enoyl-CoA. Pathway: Lipid metabolism; fatty acid beta-oxidation. Subunit structure: Homotrimer. Subcellular location: Mitochondrion matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCI
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q28
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Eci1
Gene Accession No. P23965, P42125, P42126
Gene Alias 3,2 trans-enoyl-coenyme A isomerase; 3,2 trans-enoyl-Coenzyme A isomerase; 3,2-trans-enoyl-CoA isomerase; 3,2-trans-enoyl-CoA isomerase, mitochondrial; ACBD2; acetylene-allene isomerase; D3,D2-enoyl-CoA isomerase; Dci; Delta(3),Delta(2)-enoyl-CoA isomerase; dodecenoyl-CoA de; dodecenoyl-CoA delta isomerase (3,2 trans-enoyl-CoA isomerase); dodecenoyl-CoA isomerase; dodecenoyl-Coenzyme A delta isomerase (3,2 trans-enoyl-Coenyme A isomerase); dodecenoyl-Coenzyme A delta isomerase (3,2 trans-enoyl-Coenzyme A isomerase); DRS 1; DRS1; eci; ECI1; enoyl-CoA delta isomerase 1; enoyl-CoA delta isomerase 1, mitochondrial; enoyl-Coenzyme A delta isomerase 1; HCA88; Peroxisomal 3
Gene Symbols Eci1
Host Species Rabbit
Immunogen A synthetic peptide of human DCI (aa 271-284).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13177, 1632, 29740
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.