Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dcia, Fluorescent Thiol Reactive Neutral Probe, Solid, 10 mg

Catalog No. 502705291
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5291 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5291 Supplier Abcam Supplier No. AB14532110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Self-quenching fluorescent probe that regains strong fluorescence upon reaction with thiol groups.

  • Molecular weight: 490.33 g/mol
  • Purity: >85%
  • Form: Solid
  • Properties: Fluorescent thiol-reactive neutral probe; self-quenching until reaction with thiol groups
  • Composition: 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin
  • CAS Number: 76877-34-4

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Thiol Groups
Substrate Thiol groups
Shipping Condition Ambient
Label or Dye Fluorescent Probe
Buffer 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin
Content And Storage Store at -20°C.
Form Solid
Detection Method Fluorescence
Formulation 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin
Quantity 10 mg
For Use With (Application) Thiol labeling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.