Learn More
Abcam DCIA, Fluorescent Thiol-reactive Neutral Probe, Solid, 20 mg
Description
- Chemical Type: Fluorescent thiol-reactive neutral probe
- Molecular Weight: 490.33 g/mol
- Purity: >85%
- Composition: 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin
- Physical Form: Solid
- Functional Properties: Self-quenching probe; regains strong fluorescence upon reaction with thiol groups
- Storage Temperature: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Thiol Groups |
| Shipping Condition | Ambient |
| Substrate | Thiol groups |
| Label or Dye | Fluorescent Probe |
| Form | Solid |
| Product Type | Protein Labeling Reagent |
| Buffer | 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.