Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCP1B Monoclonal Antibody (OTI2D8), TrueMAB™, OriGene
SDP

Catalog No. 501683106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3106 Supplier OriGene Supplier No. TA809932
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.

Specifications

Antigen DCP1B
Applications Western Blot
Classification Monoclonal
Clone OTI2D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DCP1B
Gene Accession No. Q8IZD4
Gene Alias DCP1
Gene Symbols DCP1B
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 167-473 of human DCP1B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196513
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.