Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCPIB, Tocris Bioscience™
GSA_VAGREENER_CHOICE

Catalog No. p-4783398
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-401-0 10 mg
15-405-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-401-0 Supplier R&D Systems Supplier No. 1540/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Blocker of VRAC/ICl, swell

Volume-regulated anion channel (VRAC) blocker (IC50 ∽ 2 μM in rat pancreatic β-cells). Also blocks ICl,swell in various cardiovascular tissues (IC50 = 4.1 μM in CPAE cells). Inhibits glucose-stimulated insulin secretion in intact β-cells via VSAC inhibition and indirect KATP channel activation. Reverses cell swelling-induced action potential duration shortening in atrial myocytes and inhibits astroglial swelling in vitro. Also activates TREK1 and TRAAK K+ channels and inhibits TRESK, TASK1 and TASK3 K+ channels at 10 μM, in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 82749-70-0
Molecular Formula C22H28Cl2O4
Molecular Weight (g/mol) 427.362
InChI Key KHKGTPJPBOQECW-UHFFFAOYSA-N
Synonym SN-401, 4-2-butyl-6,7-dichlor-2-cyclopentyl-indan-1-on-5-yl oxybutyric acid, d0j3qc, 4-2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-3h-inden-5-yl oxy butanoic acid, 4-2-butyl-6,7-dichloro-2-cyclopentylindan-1-on-5-yl oxybutyric acid, 4-2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-2,3-dihydro-1h-inden-5-yl oxy butanoic acid, 4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid, + 4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid, -4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid
PubChem CID 10071166
IUPAC Name 4-[(2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-3H-inden-5-yl)oxy]butanoic acid
SMILES CCCCC1(CC2=CC(=C(C(=C2C1=O)Cl)Cl)OCCCC(=O)O)C3CCCC3

Specifications

Quantity 10 mg
Formula Weight 427.37
Percent Purity >98%
Chemical Name or Material DCPIB
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.