Learn More
DCPS Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305444A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Necessary for the complete degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA fragments shorter than 10 nucleotides that are produced by 3′->5′ exosome-mediated mRNA decay. Releases m7GMP. Can also degrade m7GDP to m7GMP. Has no activity towards mRNA molecules longer than 25 nucleotides.Specifications
DCPS | |
Polyclonal | |
Unconjugated | |
DCPS | |
HINT-5, HSL1, DCS-1, heat shock-like protein 1, hint-related 7meGMP-directed hydrolase, histidine triad protein member 5, homolog of C. elegans 7meGMP-directed hydrolase dcs-1, mRNA decapping enzyme, scavenger mRNA-decapping enzyme DcpS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28960 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96C86 | |
DCPS | |
Between 287 and 337 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.