Learn More
Vernier™ DCU Terminal Plug Kit
SDPSCI_ED
Catalog No. S16139ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Digital Control Unit
Catalog No. For Use With (Equipment)
S16139ND Digital Control Unit
1 options
Catalog No. S16139ND Supplier Vernier™ Supplier No. DCUPLUG
Add to Cart
Add to Cart
Includes
Four detachable screw terminal plugs for the DCU, One precision slotted screwdriver
This kit contains four detachable screw terminal plug connectors for the Vernier Digital Control Unit (DCU) and one precision-slotted screwdriver.
  • The screw terminal plug connector is a detachable wiring plug for your DCU. The DCU is shipped with one plug, but it may be useful to have extras. Store your favorite projects with the electronics wired to their own plug and save time by not having to rewire them. When you are ready to pull it off the shelf, you can simply plug it into the DCU and run your project. In addition, you may want your students to be able to work on individual projects and share a DCU. Simply connect the plug when it is time to test or present each project.
  • This kit includes four detachable plug connectors and one precision screwdriver that works with the small screws of the terminal plug.
Warranty and Services
5-year limited warranty
Specifications
Includes Four plugs and one screwdriver
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Digital Control Unit
WARNING: Cancer - www.P65Warnings.ca.gov