Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729896 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9896 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9896 Supplier Proteintech Group Inc Supplier No. 149951AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in ubiquitination and subsequent proteasomal degradation of target proteins. Component of the DDD- E2 complexes which may provide a platform for interaction with CUL4A and WD repeat proteins.

Specifications

Antigen DDA1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDA1
Gene Accession No. Q9BW61, Q9D9Z5
Gene Alias C19orf58, DDA1, DET1 and DDB1 associated 1, PCIA 1, PCIA1
Gene Symbols Dda1
Host Species Rabbit
Immunogen DDA1 Fusion Protein Ag6981
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66498, 688813, 79016
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.