Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114071 Supplier Invitrogen™ Supplier No. PA5114071
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KNVLVIGTTG TQTYFLPEGE LPLCSRMVSG QDESSDKEIT HSVMDSGRKE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDI1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DDI1
Gene Accession No. Q8WTU0
Gene Alias 1700011N24Rik; DDI1; DDI1, DNA-damage inducible 1, homolog 1; DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae); DNA damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1 (S. cerevisiae); DNA-damage inducible protein 1; protein DDI1 homolog 1; RGD1559430
Gene Symbols DDI1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DDI1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414301
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.