Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ddit3 Monoclonal Antibody (9C8), Active Motif™
SDP

Catalog No. p-7188687
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3334 10 μg
50-199-3333 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3334 Supplier Active Motif Supplier No. 61350
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.

Specifications

Antigen Ddit3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene DDIT3
Gene Accession No. P35639
Gene Alias C/EBP homologous protein; C/EBP homoologous protein 10; C/EBP zeta; C/EBP-homologous protein; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; CEBPZ; CHOP; CHOP10; CHOP-10; DDIT3; DDIT-3; DNA damage inducible transcript 3; DNA damage-inducible transcript 3 protein; DNA-damage inducible transcript 3; DNA-damage-inducible transcript 3; Gadd153; Growth arrest and DNA damage-inducible protein GADD153; Growth arrest and DNA-damage-inducible protein GADD153; MGC4154
Gene Symbols DDIT3
Host Species Mouse
Immunogen This antibody was raised against full-length recombinant mouse Ddit3 protein.
Purification Method Protein G
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13198
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.