Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDIT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520495 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520495 Supplier Invitrogen™ Supplier No. PA520495
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human kidney tissue lysate. PA5-20495 can be used with blocking peptide PEP-0615.

DDIT4 gene ontology annotations related to this gene include cellular response to dexamethasone stimulus; intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator; negative regulation of glycolytic process; negative regulation of TOR signaling; protein-containing complex disassembly; reactive oxygen species metabolic process; response to hypoxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDIT4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DDIT4
Gene Accession No. Q8VHZ9, Q9D3F7, Q9NX09
Gene Alias 5830413E08Rik; AA415483; DDIT4; Dexamethasone-induced gene 2 protein; dig2; DNA damage inducible transcript 4; DNA damage-inducible transcript 4 protein; DNA-damage-inducible transcript 4; DNA-damage-inducible transcript 4 protein; HIF-1 responsive protein RTP801; HIF-1 responsive RTP801; protein regulated in development and DNA damage response 1; REDD1; REDD-1; RP11-442H21.1; RTP801
Gene Symbols DDIT4
Host Species Rabbit
Immunogen a 12 amino acid peptide from near the center of human RTP801.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140942, 54541, 74747
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.