Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDK (FLAG) Mouse anti-Tag, Magnetic beads, Polyclonal, liquid, TrueMAB™
SDP

Catalog No. 50786586
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-786-586 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-786-586 Supplier OriGene Supplier No. TA150042
Only null left
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

Synthetic peptide sequence is DYKDDDDK

DDK (FLAG-tag, FLAG octapeptide, FLAG epitope) is a polypeptide protein tag that can be added to a protein using recombit DNA technology, having the sequence motif DYKDDDDK (where D=aspartic acid, Y=tyrosine, and K=lysine). It is an artificial antigen to which specific, high affinity monoclonal antibodies have been developed and hence can be used for protein purification by affinity chromatography and also can be used for locating proteins within living cells. It has been used to separate recombit, overexpressed protein from wild-type protein expressed by the host organism. It can also be used in the isolation of protein complexes with multiple subunits, because its mild purification procedure tends not to disrupt such complexes. It has been used to obtain proteins of sufficient purity and quality to carry out 3D structure determination by x-ray crystallography.

Specifications

Antigen DDK (FLAG)
Applications Immunoprecipitation
Classification Polyclonal
Clone OTI4C5
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alias ANTI DDK, ANTI-DDK, ANTIDDK, FLAG TAG, ANTI FLAG, ANTI-FLAG, ANTI FLAG ANTIBODY, FLAG ANTIBODY, M2 ANTIBODY, FLAG M2, ANTIBODY FOR FLAG, 4C5, ANTI DDK 4C5, DDK 4C5, ANTI-DDK 4C5, DYKDDDDK TAG, D-TAG ANTIBODY, ANTI D-TAG, ANTI D TAG, ANTI-D TAG, FLAG AB, FLAG MAB, DDK ANTIBODY
Host Species Mouse
Immunogen Anti-DDK monoclonal antibody is produced by immunizing mice with a synthetic peptide coupled to KLH
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.