Learn More
Carbosynth LLC. n-Dodecyl-beta-D-maltoside, 69227-93-6, MFCD00043012, 5 g
SDP
Supplier:  Carbosynth LLC. DD061995G
Encompass_Preferred
Formula: C24H46O11, Molecular weight: 510.63 g/mol, 98%, Store at -15°C, Synonyms: Lauryl maltoside; Lauryl-beta-D-maltoside; Lauryl-beta-D-maltopyranoside.
Catalog No. 50-852-081
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.