Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557988 Supplier Invitrogen™ Supplier No. PA557988
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLIPHTYPDT PIHTQKQWFR ETFNHLFAIA NSAEAGDAGV HLVSGWQIFQ STPTEEVPFW ADVVLGFRKM TE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.

DDO is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDO
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDO
Gene Accession No. Q99489
Gene Alias 5330420D20Rik; 5730402C02Rik; AI467244; aspartic oxidase; DASOX; D-aspartate oxidase; D-aspartate oxidase, DDO; Ddo; DDO protein; DDO-1; DDO-2
Gene Symbols DDO
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDO. Recombinant protein control fragment (Product #RP-95047).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.