Learn More
Apexbio Technology LLC DDR1-IN-1 1449685-96-4 10mg
SDP
Supplier:  Apexbio Technology LLC B338310
Encompass_Preferred
DDR1-IN-1 (CAS 1449685-96-4) is a small-molecule inhibitor that selectively targets discoidin domain receptor 1 (DDR1) a receptor tyrosine kinase implicated in cell adhesion migration and extracellular matrix remodeling DDR1-IN-1 exhibits high potency against DDR1 with reported IC50 and EC50 values of 105 nM and 87 nM respectively The compound demonstrates reduced activity toward DDR2 showing a 4-fold higher IC50 (413 nM) DDR1-IN-1 is utilized in biomedical research to investigate DDR1-mediated signaling pathways and to explore the therapeutic potential of DDR1 inhibition in disease models involving aberrant receptor activation
Catalog No. 50-000-94418
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.