Learn More
Promega DDR1-NanoLuc Fusion Vector, 20microg
Catalog No. PRN2451 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRN2451 20 μg
1 options
This item is not returnable. View return policy
Catalog No. PRN2451 Supplier Promega Supplier No. N2451
This item is not returnable. View return policy
The NanoLuc Kinase Fusion Vectors contain the coding region of the indicated human kinase fused to either the C- or the N-terminus of NanoLuc luciferase. Designed for use with the NanoBRET Target Engagement (TE) Intracellular Kinase Assay.
  • Kinase Fusion Vector for NanoBRET™ Target Engagement
  • Transfection−ready DNA to express full−length DDR1 kinase fused to NanoLuc(R) luciferase
  • Measures kinase target engagement in live cells
  • Supplied as a control vector with NanoBRET™ TE Intracellular Kinase Assay, K4
Specifications
Content And Storage -30°C to -10°C
Quantity 20 μg