Learn More
Description
Recommended positive controls: HeLa, NIH-3T3, JC. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (86%), Rhesus Monkey (100%), Bovine (100%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | DDX23 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Ddx23 |
| Gene Accession No. | Q9BUQ8 |
| Gene Alias | 100 kDa U5 snRNP-specific protein; 3110082M05Rik; 4921506D17Rik; DDX23; DEAD (Asp-Glu-Ala-Asp) box polypeptide 23; DEAD box protein 23; DEAD-box helicase 23; probable ATP-dependent RNA helicase DDX23; prp28; PRP28 homolog; PRP28 homolog, yeast; PRP28p homolog; PRPF28; SNRNP100; U5 snRNP 100 kD protein; U5-100K; U5-100KD |
| Gene Symbols | Ddx23 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
