Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX24 Monoclonal Antibody (A3B11)
GREENER_CHOICE

Catalog No. PIMA536116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536116 Supplier Invitrogen™ Supplier No. MA536116
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDX24 Monoclonal Antibody for IHC (P)

METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX24
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A3B11
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Ddx24
Gene Accession No. 0
Gene Alias 1700055J08Rik; 2510027P10Rik; AI649272; ATP-dependent RNA helicase DDX24; DDX24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; DEAD-box helicase 24; LOC553462 protein; S. cerevisiae CHL1-like helicase; si:ch1073-416d2.2
Gene Symbols Ddx24
Host Species Mouse
Immunogen Recombinant protein within zebrafish ddx24 aa 562-819.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 553462
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.