Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541281 Supplier Thermo Scientific Supplier No. PA541281
Only null left
Add to Cart
Add to Cart

DDX27 Polyclonal Antibody

DDX27 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX27
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DDX27
Gene Accession No. Q96GQ7
Gene Alias C86129; cPERP-F; Ddx27; DEAD (Asp-Glu-Ala-Asp) box polypeptide 27; DEAD box protein 27; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 27; DEAD-box helicase 27; Deficiency of ribosomal subunits protein 1 homolog; dJ686N3.1; DRS1; Drs1p; HSPC259; LOW QUALITY PROTEIN: probable ATP-dependent RNA helicase DDX27; PP3241; Probable ATP-dependent RNA helicase DDX27; probable ATP-dependent RNA helicase DDX27-like; RHLP; RNA helicase-like protein; RNA-helicase like protein; Rrp3p
Gene Symbols DDX27
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DDX27 (aa 118-167).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55661
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.