Learn More
DDX46 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
Specifications
Specifications
| Antigen | DDX46 |
| Applications | Immunohistochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | DDX46 |
| Gene Accession No. | Q569Z5, Q7L014 |
| Gene Alias | PRPF5, Prp5, DEAD box protein 46, PRP5 homolog, Prp5-like DEAD-box protein, RNA helicase, probable ATP-dependent RNA helicase DDX46 |
| Gene Symbols | Ddx46 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.