Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX5 Monoclonal Antibody (Ab204), Alexa Fluor™ 647, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 51600542
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-600-542 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-600-542 Supplier Invitrogen™ Supplier No. 51600542
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This Ab204 monoclonal antibody detects human DDX5 protein also known as DEAD box protein 5 and RNA helicase p68. This antibody has been reported to cross-react with mouse and rat orthologs of DDX5 protein. Applications Reported: This Ab204 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This Ab204 antibody has been pre-diluted and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins located at www.thermofisher.com/flowprotocols. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Applications Reported: This Ab204 antibody has been reported for use in immunocytochemical staining. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a RNA-dependent ATPase, and also a proliferation-associated nuclear antigen, specifically reacting with the simian virus 40 tumor antigen. This gene consists of 13 exons, and alternatively spliced transcripts containing several intron sequences have been detected, but no isoforms encoded by these transcripts have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX5
Applications Flow Cytometry
Classification Monoclonal
Clone Ab204
Concentration 5 μL/Test
Conjugate Alexa Fluor 647
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene DDX5
Gene Accession No. P17844
Gene Alias 2600009A06Rik; ATP-dependent RNA helicase DDX5; DDX5; ddx5 gene protein; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 5; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 5; DEAD (Asp-Glu-Ala-Asp) box helicase 5; DEAD (Asp-Glu-Ala-Asp) box polypeptide 5; DEAD box protein 5; DEAD box RNA helicase DEAD1; DEAD box-5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase, 68kD); DEAD-box helicase 5; DKFZp434E109; DKFZp686J01190; G17P1; HELR; Hlr1; HUMP68; mDEAD1; MGC118083; p68; p68 RNA helicase; PCR product with interspecies-compatible primers; probable ATP-dependent RNA helicase DDX5; RNA helicase p68; RP23-247J12.4
Gene Symbols DDX5
Host Species Mouse
Immunogen Purified T antigen-related protein from Ad2+ ND2 virus-infected HeLa cells.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1655
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.