Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541295 Supplier Invitrogen™ Supplier No. PA541295
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GPNRGAKRRR EEARQRDQEF YIPYRPKDFD SERGLSISGE GGAFEQQAAG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 92%; Rat: 93%; Zebrafish: 92%.

This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX54
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DDX54
Gene Accession No. Q8TDD1
Gene Alias 2410015A15Rik; 5-Apr; AI414901; ATP-dependent RNA helicase; ATP-dependent RNA helicase DDX54; ATP-dependent RNA helicase DP97; chunp6913; Ddx54; DEAD (Asp-Glu-Ala-Asp) box polypeptide 54; DEAD box helicase 97 KDa; DEAD box protein 54; DEAD box RNA helicase 97 kDa; DEAD-box helicase 54; DP97; LOW QUALITY PROTEIN: ATP-dependent RNA helicase DDX54; mgc2835; RGD1562539; zgc:111908
Gene Symbols DDX54
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DDX54 (aa 661-710).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79039
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.