Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva DEAE-Sepharose™ Media

Catalog No. 45000134 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-134 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-134 Supplier Cytiva Supplier No. 17071001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Developed for process-scale chromatography

  • Weak anion exchanger with excellent flow properties
  • High capacity for proteins of all pI values
  • Ion exchange group is diethylaminoethyl, which remains charged and maintains consistently high capacities over the entire working range, pH 3 to 9

TRUSTED_SUSTAINABILITY

Specifications

Buffer 20mM N-methylpiperazine, 20mM Piperazine, 20mM L-Histidine, 20mM Bis-Tris, 20mM Bis-Tris Propane, 20mM Triethanolamine, 20mM Tris, 50mM N-methyldiethanolamine, 20mM Diethanolamine at pH 8.4, 20mM 1,3-Diaminopropane, 20mM Ethanolamine, 20mM Piperazine, 20mM 1,3-Diaminopropionate
Product Type DEAE Sepharose CL-6B media
Quantity 500 mL
Flow Rate <150cm/hr.
Ligand Type Diethylaminoethyl
pH Range 3 to 9
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) Process scale chromato graphy
Matrix DEAE-Sepharose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.