Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Deaza dGTP

Catalog No. 45001348 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-348 2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-348 Supplier Cytiva Supplier No. 27209002
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reduce band compressions in sequencing gels. Deaza dGTP uses nucleotide analogues in the sequencing mixes to reduce compressions.

Band compressions are ambiguous band patterns on sequencing gels caused by stable intra-strand secondary structures which are not fully denatured during electrophoresis. Their occurrence can be reduced by substituting nucleotide analogues in the sequencing mixes. To resolve G-band compressions, use 7-deaza dGTP (cd7GTP).

Order Info

Supplied as convenient solutions and supplied in purified water as 5 mM solutions.

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7 and 7.5
Concentration 5 mM
Quantity 2 μm
Color Colorless
For Use With (Application) Reduces b and compression in sequencing Gels
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.