Learn More
MilliporeSigma™ Supelco™ Deburring Tool configured for 1/8 in.. Tubing
Catalog No. 111009955 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-9955 1/Pk.
1 options
Catalog No. 11-100-9955 Supplier MilliporeSigma™ Supelco™ Supplier No. 58806
Deburring tool is an accessory that can be used with 1/8 in. tubing for dressing it without distortion after being smooth-cut using the tubing cutter.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.