Learn More
Strem, An Ascensus Company DECABORANE 1G
SDP
Supplier:  Strem, An Ascensus Company 0505001G
Encompass_Preferred
DECABORANE, 97% 1g CAS#17702-41-9 MFCD00011314 B₁₀H₁₄ FW:122.21
Catalog No. NC2167412
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.