Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™

Catalog No. AC406170010
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406170010 1 L AcroSeal™ Glass Bottle
AC406171000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC406170010 Supplier Thermo Scientific Chemicals Supplier No. 406170010
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-17-8 | C10H18 | 138.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Melting Point -31.0°C
Density 0.8900g/mL
Boiling Point 187.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (mixture of cis and trans) (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4740 to 1.4760
Linear Formula C10H18
Quantity 1 L
Beilstein 05,III,245
Merck Index 15,285
Specific Gravity 0.89
Solubility Information Solubility in water: slightly soluble (6mg/l at 20°C). Other solubilities: very soluble in acetone,benzene,ether and,ethanol
Viscosity 3 mPa.s (20°C)
Formula Weight 138.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Decahydronaphthalene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 202-046-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.