Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decahydronaphthalene, Spectrum™ Chemical
SDP

Catalog No. 18620508 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-508 500 mL
18-620-507 100 mL
18-620-510 2.5 L
18-620-509 6 x 500 mL
18-620-511 4 x 2.5 L
18-620-512 20 L
18-620-513 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-620-508 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1000500MLGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 91-17-8

Specifications

Boiling Point 170.6°C
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 97%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.