Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decahydronaphthalene, Spectrum™ Chemical
SDP

Catalog No. 18620513 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-513 200 L
18-620-507 100 mL
18-620-508 500 mL
18-620-510 2.5 L
18-620-509 6 x 500 mL
18-620-511 4 x 2.5 L
18-620-512 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-620-513 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1000200LTBL

Chemical Identifiers

CAS 91-17-8

Specifications

Boiling Point 170.6°C
Packaging Drum
Quantity 200 L
Percent Purity 97%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.