Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decamethylferrocene, 99%, Thermo Scientific Chemicals

Catalog No. p-7050757
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39406MD 250 mg
AA3940603 1 g
AA3940606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA39406MD Supplier Thermo Scientific Chemicals Supplier No. 039406MD
Only null left
Add to Cart

Decamethylferrocene is used as a superior internal reference redox standard to ferrocene in voltammetric and amperometric titrations. It is used as photocathode for the detection of barium fluoride scintillation light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12126-50-0
Molecular Formula C20H30Fe
Molecular Weight (g/mol) 326.31
MDL Number MFCD00058736
InChI Key VKRKSGODPDKGAD-UHFFFAOYSA-N
Synonym Bis(pentamethylcyclopentadienyl)iron(II)
SMILES Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Fe]2345

Specifications

Melting Point 298°C to 300°C (sublimation)
Odor Odorless
Quantity 250 mg
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 326.31
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Decamethylferrocene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.