Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decanenitrile 98.0+%, TCI America™
SDP

Catalog No. D2426100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2426100ML 100 mL
D242625ML 25 mL
D2426500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2426100ML Supplier TCI America Supplier No. D2426100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1975-78-6
Molecular Formula C10H19N
Molecular Weight (g/mol) 153.269
MDL Number MFCD00039505
InChI Key HBZDPWBWBJMYRY-UHFFFAOYSA-N
Synonym decanonitrile, caprinitrile, 1-cyanononane, n-decanenitrile, nonyl cyanide, n-nonyl cyanide, unii-1602vq5ivc, 1-decanenitrile, nitrile 10 d, decanoic acid nitrile
PubChem CID 74791
IUPAC Name decanenitrile
SMILES CCCCCCCCCC#N

Specifications

Melting Point -15°C
Color Yellow
Boiling Point 237°C
Quantity 100 mL
Formula Weight 153.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decanenitrile
EINECSNumber (2)-1507
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.