Learn More
Medchemexpress LLC Decanoic acid-13C | 84600-66-8 | MFCD00083932 | 98.0% | 173.26 g·mol⁻¹ | C9[13C]H20O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW26618825MG
Encompass_Preferred
Decanoic acid-13C is a carbon-13 isotopically labeled decanoic (capric) acid supplied as a solid for use as a tracer or internal standard in analytical and research workflows. It is intended for NMR, GC-MS, and LC-MS applications and is provided with high reported purity.
  • Isotopically labeled decanoic acid for tracer and internal standard use.
  • High purity (98.0%) appropriate for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Solid, white to off-white appearance for convenient handling.
  • Storage guidance: stable at room temperature as solid; store solutions at -20°C to -80°C as recommended.
Catalog No. 50-002-81454