Learn More
Medchemexpress LLC Decanoic acid-13C | 84600-66-8 | MFCD00083932 | 98.0% | 173.2573 g/mol | C10H20O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW26618850MG
Encompass_Preferred
Decanoic acid-13C is a stable isotope-labeled decanoic (capric) acid with a 13C label, supplied for research use in metabolic tracing and analytical applications. CAS number 84600-66-8; reported purity 98.0%.
  • Stable isotope-labeled decanoic acid (13C) for tracer studies.
  • High purity (98.0%) suitable for analytical work.
  • Useful as an internal standard for mass spectrometry.
  • Available in small research pack sizes (25 mg-5 g).
  • Data sheet and safety data sheet available.
Catalog No. 50-002-80687