Learn More
Sigma Aldrich Fine Chemicals Biosciences Cinnamaldehyde natural 955KG
Supplier: Sigma Aldrich Fine Chemicals Biosciences W2286135KGK
Cinnamaldehyde is an -unsaturated aromatic aldehyde that can be used as a flavoring agent. It is the principal flavor component of cinnamon oil.