Learn More
Medchemexpress LLC Decanoic acid-d19 | 88170-22-3 | 98.4% | 191.38 g/mol | C10HD19O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015309S1100MG
Encompass_Preferred
Decanoic acid-d19 is the deuterium-labeled analogue of decanoic acid supplied for research and analytical applications. The isotopically labeled fatty acid enables tracing and mass spectrometry studies, and is provided as a high-purity material suitable for mechanistic, pharmacological, and analytical research.
  • Deuterium-labeled decanoic acid for isotope tracing and MS analyses.
  • High purity (98.4%) suitable for analytical applications.
  • Provided in a 100 mg package for laboratory use.
  • Useful for studies of ionotropic glutamate receptor modulation and seizure-related research.
  • CAS number 88170-22-3; molecular weight 191.38 g/mol for reference.
Catalog No. 50-003-03654