Learn More
Medchemexpress LLC Decanoic acid-d19 | 88170-22-3 | 98.4% | 191.38 | C10HD19O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW015309S15MG
Encompass_Preferred
Decanoic acid-d19 is the deuterium-labeled form of decanoic (capric) acid used as an isotope-labeled standard and tracer in analytical, metabolic, and pharmacokinetic research. It is a deuterated fatty acid reported to be brain-penetrant and a non-competitive AMPA receptor inhibitor; the material is supplied for laboratory use with batch documentation available.
  • Deuterium-labeled (d19) analog for isotope tracing.
  • High reported purity: 98.4%.
  • Molecular formula C10HD19O2; molecular weight 191.38.
  • Supplied in small pack sizes suitable for analytical work.
  • Provided with certificate of analysis and safety data sheet.
  • Suitable for metabolic, pharmacokinetic, and tracer studies.
  • Useful as an internal standard for mass spectrometry.
Catalog No. 50-003-03608