Learn More
Medchemexpress LLC Decanoic acid-d3 | 102611-15-4 | MFCD00144050 | ≥99.0% | 175.28 | C10H17D3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW015309S
Encompass_Preferred
Decanoic acid-d3 is the deuterium-labeled analogue of decanoic (capric) acid supplied as a white to off-white solid for research use. It is used as an isotopically labeled standard for tracing, quantification, and mechanistic studies. Decanoic acid is brain-penetrant and acts as a non-competitive AMPA receptor inhibitor with reported antiseizure activity.
  • High isotopic purity (≥99.0%).
  • Suitable as an internal standard for LC/GC and metabolic tracing.
  • White to off-white solid, stable under recommended storage.
  • Storage: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes for analytical use.
Catalog No. 50-003-02232