Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Decitabine
GREENER_CHOICE

Catalog No. 262410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-241-0 10 mg
26-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-241-0 Supplier Tocris Bioscience™ Supplier No. 2624/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

DNA methyltransferase inhibitor

Cytosine analog that once incorporated into DNA acts as a suicide substrate for DNA methyltransferase. Inhibits DNA methyltransferase and results in DNA hypomethylation and activation of silent genes. Chemotherapeutic agent; suppresses growth of human tumor cell lines. Demethylates differentiation-related genes; reverses embryonic stem cell differentiation.

Chemical Identifiers

CAS 2353-33-5
Molecular Formula C9H13N3O4

Specifications

Quantity 10 mg
Formula Weight 228.21
Purity >99%
Molecular Weight (g/mol) 227.22
Chemical Name or Material Decitabine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.