Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-CYCLE™ II Germicidal Detergent

Catalog No. 19546598 Shop All VAI Products
Change view
Click to view available options
Dilution:
1:128
1:256
Volume (Metric):
473 mL
3.8 L
Format:
Concentrate
SimpleMix
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dilution Volume (Metric) Format
19-546-598 1:128 - -
19-546-601 - 473 mL SimpleMix
19-546-528 1:256 473 mL SimpleMix
19-546-597 1:256 3.8 L Concentrate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-546-598 Supplier VAI Supplier No. DCY2041Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report

DECON-CYCLE™ II Germicidal Detergent is a low pH phenolic that is effective as hard non-porous surface disinfectant/detergent formulated for effective aseptic and controlled environment sanitation.

A low pH, dilutable, hospital detergent effective in 10 minutes at 20°C in hard water up to 400 ppm (calculated as CaCO3) in the presence of 5% blood serum. Recommended for use in pharmaceutical, biotechnology, medical device manufacturing, and in healthcare institutions to control cross-contamination hazards.

  • Multi-phenolic formula designed to clean, disinfect, and deodorize any washable inanimate surface.
  • Filtered at 0.2 μm and gamma irradiated.
  • Strong enough to kill a broad spectrum of pathogenic bacteria, yet mild enough to have no harmful effect on disinfected surfaces.
  • Completely traceable and validated for sterility and shelf life.
  • Filled in an ISO 5 (Grade A/B, Former Class 100).
  • Lot sterility tested according to current USP compendium.
  • Completely traceable from start to finish.

Order Info

Double-bag packaging.

Compliance

Completely tested according to current USP compendium.

Specifications

Sterility Sterile
Dilution 1:128
For Use With (Application) Kills broad spectrum of pathogenic Bacteria, pharmaceutical, biotechnology, medical device and health care industries
Volume (English) 1 gal.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.