Learn More
Description
Designed for use where a broad-spectrum disinfectant is required in moderate amounts of organic soil such as decontamination of walls, ceilings and floors.
- Filtered at 0.2μm and gamma irradiated to a 10-6 Sterility Assurance Level
- Strong enough to kill a broad spectrum of pathogenic and non-pathogenic bacteria; mild enough to have no harmful effect on disinfected surfaces
- Detergent that has efficacy performance that confirms to AOAC protocol testing at a use dilution of 1:128 in he presence of 5% blood serum at 20°C with an exposure time of 10 minutes
- Completely traceable and validated for sterility and shelf life
Order Info
Double-bagged packaged
Certifications
Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
Compliance
Completely tested according to current USP compendium
Specifications
Specifications
| Form | Concentrate |
| Sterility | Sterile |
| Certifications/Compliance | ISO 5 |
| For Use With (Application) | For use by pharmaceutical and biotechnology industries, medical device manufacturers, hospitals and any healthcare institutions |
| Volume (English) | 1 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.