Learn More
VAI DECON-PHENE™ Germicidal Detergent
Catalog No. 19547041 Shop All VAI Products
Change view
Click to view available options
Volume (English):
4 oz.
16 oz.
1 gal.
Sterility:
Non-sterile
Sterile
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility
19-547-041 16 oz. Sterile
19-547-038 1 gal. Sterile
19-546-846 4 oz. Sterile
19-547-037 1 gal. Sterile
19-547-042 16 oz. Non-sterile
19-546-844 1 gal. Sterile
19-547-043 16 oz. Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-547-041 Supplier VAI Supplier No. DP0616Z02
Sodium-based, phosphate-free phenolic synthetic germicidal detergent
Designed for use where a broad-spectrum disinfectant is required in moderate amounts of organic soil such as decontamination of walls, ceilings and floors.
  • Filtered at 0.2μm and gamma irradiated to a 10-6 Sterility Assurance Level
  • Strong enough to kill a broad spectrum of pathogenic and non-pathogenic bacteria; mild enough to have no harmful effect on disinfected surfaces
  • Detergent that has efficacy performance that confirms to AOAC protocol testing at a use dilution of 1:128 in he presence of 5% blood serum at 20°C with an exposure time of 10 minutes
  • Completely traceable and validated for sterility and shelf life
Order Info
Double-bagged packaged
Certifications
Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
Compliance
Completely tested according to current USP compendium
Specifications
Sterility Sterile
Certifications/Compliance ISO 5
Format SimpleMix
For Use With (Application) For use by pharmaceutical and biotechnology industries, medical device manufacturers, hospitals and any healthcare institutions
Volume (English) 16 oz.