Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-PHENE™ Germicidal Detergent

Catalog No. 19547043 Shop All VAI Products
Change view
Click to view available options
Volume (English):
4 oz.
16 oz.
1 gal.
Sterility:
Non-sterile
Sterile
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility
19-547-043 16 oz. Non-sterile
19-547-038 1 gal. Sterile
19-547-041 16 oz. Sterile
19-546-846 4 oz. Sterile
19-547-037 1 gal. Sterile
19-547-042 16 oz. Non-sterile
19-546-844 1 gal. Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-547-043 Supplier VAI Supplier No. DP0716Z02

Sodium-based, phosphate-free phenolic synthetic germicidal detergent

Designed for use where a broad-spectrum disinfectant is required in moderate amounts of organic soil such as decontamination of walls, ceilings and floors.

  • Filtered at 0.2μm and gamma irradiated to a 10-6 Sterility Assurance Level
  • Strong enough to kill a broad spectrum of pathogenic and non-pathogenic bacteria; mild enough to have no harmful effect on disinfected surfaces
  • Detergent that has efficacy performance that confirms to AOAC protocol testing at a use dilution of 1:128 in he presence of 5% blood serum at 20°C with an exposure time of 10 minutes
  • Completely traceable and validated for sterility and shelf life

Order Info

Double-bagged packaged

Certifications

Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report

Compliance

Completely tested according to current USP compendium

Specifications

Sterility Non-sterile
Certifications/Compliance ISO 5
Filtered 0.2 μm
Format SimpleMix
For Use With (Application) For use by pharmaceutical and biotechnology industries, medical device manufacturers, hospitals and any healthcare institutions
Volume (English) 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.